1-(2-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid, tech

Lieferant: Apollo Scientific

PC28082-1G
APOSPC28082-1GEA 1570 EUR
APOSPC28082-1G
1-(2-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid, tech
1-(2-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₂H₁₁FN₂O₂
Molecular Weight: 234,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07021455
CAS-Nummer: 956930-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR