2-[4-(4-Fluorophenyl)piperazin-1-yl]-2-methylpropanenitrile, tech

Lieferant: Apollo Scientific

PC28084-1G
APOSPC28084-1GEA 1570 EUR
APOSPC28084-1G
2-[4-(4-Fluorophenyl)piperazin-1-yl]-2-methylpropanenitrile, tech
2-[4-(4-Fluorophenyl)piperazin-1-yl]-2-methylpropanenitrile
Formel: C₁₄H₁₈FN₃
Molecular Weight: 247,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00202053
CAS-Nummer: 338754-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR