1-(4-Methylphenyl)-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one, tech

Lieferant: Apollo Scientific

PC28085-1G
APOSPC28085-1GEA 1570 EUR
APOSPC28085-1G
1-(4-Methylphenyl)-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one, tech
1-(4-Methylphenyl)-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one
Formel: C₁₁H₉F₃N₂O
Molecular Weight: 242,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215510
CAS-Nummer: 63695-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR