3-(Pyridin-3-yloxy)-5-(trifluoromethyl)pyridine-2-carbonitrile, tech

Lieferant: Apollo Scientific

PC28086-1G
APOSPC28086-1GEA 1570 EUR
APOSPC28086-1G
3-(Pyridin-3-yloxy)-5-(trifluoromethyl)pyridine-2-carbonitrile, tech
3-(Pyridin-3-yloxy)-5-(trifluoromethyl)pyridine-2-carbonitrile
Formel: C₁₂H₆F₃N₃O
Molecular Weight: 265,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664105
CAS-Nummer: 338758-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR