(2E)-3-(Dimethylamino)-1-(2-fluorophenyl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

Synonyme: (E)-3-(dimethylamino)-1-(2-fluorophenyl)prop-2-en-1-one

PC28088-1G
APOSPC28088-1GEA 1570 EUR
APOSPC28088-1G
(2E)-3-(Dimethylamino)-1-(2-fluorophenyl)prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-(2-fluorophenyl)prop-2-en-1-one
Formel: C₁₁H₁₂FNO
Molecular Weight: 193,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172154
CAS-Nummer: 138716-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR