3-Chloro-2-(pyridine-3-carbonyl)-5-(trifluoromethyl)pyridine, tech

Lieferant: Apollo Scientific

PC28089-1G
APOSPC28089-1GEA 1570 EUR
APOSPC28089-1G
3-Chloro-2-(pyridine-3-carbonyl)-5-(trifluoromethyl)pyridine, tech
3-Chloro-2-(pyridine-3-carbonyl)-5-(trifluoromethyl)pyridine
Formel: C₁₂H₆ClF₃N₂O
Molecular Weight: 286,637 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00244682
CAS-Nummer: 338953-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR