(2Z)-3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]prop-2-enal, tech
Lieferant: Apollo Scientific
PC28090-1G
APOSPC28090-1GEA
1570
EUR
APOSPC28090-1G
(2Z)-3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]prop-2-enal, tech
(2Z)-3-(Dimethylamino)-2-[3-(trifluoromethyl)phenyl]prop-2-enal
Formel:
C₁₂H₁₂F₃NO Molecular Weight: 243,225 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00232111 CAS-Nummer: 163563-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...