(3E)-3-{[(4-Fluorophenyl)methoxy]imino}propanenitrile, tech
Lieferant: Apollo Scientific
PC28091-1G
APOSPC28091-1GEA
1570
EUR
APOSPC28091-1G
(3E)-3-{[(4-Fluorophenyl)methoxy]imino}propanenitrile, tech
(3E)-3-{[(4-Fluorophenyl)methoxy]imino}propanenitrile
Formel:
C₁₀H₉FN₂O Molecular Weight: 192,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00663730 CAS-Nummer: 338960-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...