(3Z)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]piperidin-2-one, tech
Lieferant: Apollo Scientific
PC28092-1G
APOSPC28092-1GEA
1570
EUR
APOSPC28092-1G
(3Z)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]piperidin-2-one, tech
(3Z)-3-[2-(4-Fluorophenyl)hydrazin-1-ylidene]piperidin-2-one
Formel:
C₁₁H₁₂FN₃O Molecular Weight: 221,231 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00794670 CAS-Nummer: 1086239-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...