4-Chloro-2-(4-fluorophenyl)-5-(propylamino)-2,3-dihydropyridazin-3-one, tech

Lieferant: Apollo Scientific

PC28093-1G
APOSPC28093-1GEA 1570 EUR
APOSPC28093-1G
4-Chloro-2-(4-fluorophenyl)-5-(propylamino)-2,3-dihydropyridazin-3-one, tech
4-Chloro-2-(4-fluorophenyl)-5-(propylamino)-2,3-dihydropyridazin-3-one
Formel: C₁₃H₁₃ClFN₃O
Molecular Weight: 281,713 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02083241
CAS-Nummer: 478049-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR