4-Chloro-2-(4-fluorophenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech

Lieferant: Apollo Scientific

PC28094-1G
APOSPC28094-1GEA 1570 EUR
APOSPC28094-1G
4-Chloro-2-(4-fluorophenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech
4-Chloro-2-(4-fluorophenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one
Formel: C₁₃H₁₁ClFN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD02083242
CAS-Nummer: 478049-16-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR