3-{2-[3-(Trifluoromethyl)phenyl]hydrazin-1-ylidene}pentane-2,4-dione, tech

Lieferant: Apollo Scientific

PC28095-1G
APOSPC28095-1GEA 1570 EUR
APOSPC28095-1G
3-{2-[3-(Trifluoromethyl)phenyl]hydrazin-1-ylidene}pentane-2,4-dione, tech
3-{2-[3-(Trifluoromethyl)phenyl]hydrazin-1-ylidene}pentane-2,4-dione
Formel: C₁₂H₁₁F₃N₂O₂
Molecular Weight: 272,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315662
CAS-Nummer: 58534-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR