1,1,1-Trifluoro-3-[(1-phenylethyl)amino]propan-2-ol, tech

Lieferant: Apollo Scientific

PC28098-1G
APOSPC28098-1GEA 1570 EUR
APOSPC28098-1G
1,1,1-Trifluoro-3-[(1-phenylethyl)amino]propan-2-ol, tech
1,1,1-Trifluoro-3-[(1-phenylethyl)amino]propan-2-ol
Formel: C₁₁H₁₄F₃NO
Molecular Weight: 233,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02083024
CAS-Nummer: 478050-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR