3-(4-Benzylpiperazin-1-yl)-1,1,1-trifluoropropan-2-ol, tech

Lieferant: Apollo Scientific

PC28099-1G
APOSPC28099-1GEA 1570 EUR
APOSPC28099-1G
3-(4-Benzylpiperazin-1-yl)-1,1,1-trifluoropropan-2-ol, tech
3-(4-Benzylpiperazin-1-yl)-1,1,1-trifluoropropan-2-ol
Formel: C₁₄H₁₉F₃N₂O
Molecular Weight: 288,309 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02083025
CAS-Nummer: 478050-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR