1,1,1-Trifluoro-3-(1,2,3,4-tetrahydroisoquinolin-2-yl)propan-2-ol, tech

Lieferant: Apollo Scientific

PC28100-1G
APOSPC28100-1GEA 1570 EUR
APOSPC28100-1G
1,1,1-Trifluoro-3-(1,2,3,4-tetrahydroisoquinolin-2-yl)propan-2-ol, tech
1,1,1-Trifluoro-3-(1,2,3,4-tetrahydroisoquinolin-2-yl)propan-2-ol
Formel: C₁₂H₁₄F₃NO
Molecular Weight: 245.241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02083027
CAS-Nummer: 478050-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR