(3E)-4-[Benzyl(methyl)amino]-1,1,1-trifluorobut-3-en-2-one, tech

Lieferant: Apollo Scientific

PC28102-1G
APOSPC28102-1GEA 1570 EUR
APOSPC28102-1G
(3E)-4-[Benzyl(methyl)amino]-1,1,1-trifluorobut-3-en-2-one, tech
(3E)-4-[Benzyl(methyl)amino]-1,1,1-trifluorobut-3-en-2-one
Formel: C₁₂H₁₂F₃NO
Molecular Weight: 243,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02661703
CAS-Nummer: 1164486-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR