1,1,1-Trifluoro-4-(phenylamino)butan-2-ol, tech

Lieferant: Apollo Scientific

PC28103-1G
APOSPC28103-1GEA 1570 EUR
APOSPC28103-1G
1,1,1-Trifluoro-4-(phenylamino)butan-2-ol, tech
1,1,1-Trifluoro-4-(phenylamino)butan-2-ol
Formel: C₁₀H₁₂F₃NO
Molecular Weight: 219,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012806
CAS-Nummer: 439109-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR