8-Fluoro-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 95%

Lieferant: Apollo Scientific

PC28107-1G
APOSPC28107-1GEA 224 EUR
APOSPC28107-1G
8-Fluoro-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 95%
8-Fluoro-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole
Formel: C₁₁H₁₁FN₂
Molecular Weight: 190.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00277692
CAS-Nummer: 39876-39-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR