Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylate, tech

Lieferant: Apollo Scientific

PC28108-1G
APOSPC28108-1GEA 1570 EUR
APOSPC28108-1G
Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylate, tech
Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylate
Formel: C₁₅H₁₁FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05256131
CAS-Nummer: 866041-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR