4-[4-Nitro-2-(trifluoromethyl)phenyl]morpholine 97%

Lieferant: Apollo Scientific

Synonyme: 4-(4-Nitro-2-(trifluoromethyl)phenyl)morpholine

PC28109-5G
APOSPC28109-5GEA 180 EUR
APOSPC28109-5G
4-[4-Nitro-2-(trifluoromethyl)phenyl]morpholine 97%
4-[4-Nitro-2-(trifluoromethyl)phenyl]morpholine
Formel: C₁₁H₁₁F₃N₂O₃
Molecular Weight: 276,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00100093
CAS-Nummer: 54672-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR