Methyl 2,2-difluoro-2-(4-fluorophenoxy)acetate, tech
Lieferant: Apollo Scientific
PC28111-1G
APOSPC28111-1GEA
1570
EUR
APOSPC28111-1G
Methyl 2,2-difluoro-2-(4-fluorophenoxy)acetate, tech
Formel:
C₉H₇F₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173165 CAS-Nummer: 338792-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...