Ethyl 5,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, tech

Lieferant: Apollo Scientific

PC28114-1G
APOSPC28114-1GEA 1120 EUR
APOSPC28114-1G
Ethyl 5,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, tech
Ethyl 5,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Formel: C₁₂H₉F₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00109441
CAS-Nummer: 185011-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR