Ethyl 6-chloro-8-fluoro-4-hydroxyquinoline-3-carboxylate, tech
Lieferant: Apollo Scientific
PC28115-1G
APOSPC28115-1GEA
1570
EUR
APOSPC28115-1G
Ethyl 6-chloro-8-fluoro-4-hydroxyquinoline-3-carboxylate, tech
Ethyl 6-chloro-8-fluoro-4-hydroxyquinoline-3-carboxylate
Formel:
C₁₂H₉ClFNO₃ Molecular Weight: 269,656 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173397 CAS-Nummer: 338795-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...