Ethyl 8-chloro-6-fluoro-4-hydroxyquinoline-3-carboxylate, tech

Lieferant: Apollo Scientific

PC28116-1G
APOSPC28116-1GEA 1570 EUR
APOSPC28116-1G
Ethyl 8-chloro-6-fluoro-4-hydroxyquinoline-3-carboxylate, tech
Ethyl 8-chloro-6-fluoro-4-hydroxyquinoline-3-carboxylate
Formel: C₁₂H₉ClFNO₃
Molecular Weight: 269,656 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173402
CAS-Nummer: 338795-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR