1-Methyl-2-oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

PC28117-1G
APOSPC28117-1GEA 1570 EUR
APOSPC28117-1G
1-Methyl-2-oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech
1-Methyl-2-oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₄H₉F₃N₂O
Molecular Weight: 278,229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00232375
CAS-Nummer: 338964-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR