N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-N'-methylprop-2-enehydrazide, tech

Lieferant: Apollo Scientific

PC28118-1G
APOSPC28118-1GEA 1570 EUR
APOSPC28118-1G
N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-N'-methylprop-2-enehydrazide, tech
N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-N'-methylprop-2-enehydrazide
Formel: C₁₀H₉ClF₃N₃O
Molecular Weight: 279,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01443836
CAS-Nummer: 338965-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR