2-[(2-Fluorophenyl)methylidene]propanedinitrile, tech

Lieferant: Apollo Scientific

PC28119-1G
APOSPC28119-1GEA 1570 EUR
APOSPC28119-1G
2-[(2-Fluorophenyl)methylidene]propanedinitrile, tech
2-[(2-Fluorophenyl)methylidene]propanedinitrile
Formel: C₁₀H₅FN₂
Molecular Weight: 172,159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00224065
CAS-Nummer: 2698-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR