2-[(2-Fluorophenyl)methyl]propanedinitrile, tech

Lieferant: Apollo Scientific

PC28120-1G
APOSPC28120-1GEA 1570 EUR
APOSPC28120-1G
2-[(2-Fluorophenyl)methyl]propanedinitrile, tech
2-[(2-Fluorophenyl)methyl]propanedinitrile
Formel: C₁₀H₇FN₂
Molecular Weight: 174,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231635
CAS-Nummer: 338965-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR