1-[(4-Fluorophenyl)methoxy]-1H-imidazole-2-carbaldehyde, tech

Lieferant: Apollo Scientific

PC28121-1G
APOSPC28121-1GEA 1570 EUR
APOSPC28121-1G
1-[(4-Fluorophenyl)methoxy]-1H-imidazole-2-carbaldehyde, tech
1-[(4-Fluorophenyl)methoxy]-1H-imidazole-2-carbaldehyde
Formel: C₁₁H₉FN₂O₂
Molecular Weight: 220,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871563
CAS-Nummer: 551921-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR