3-(2,3-Dihydro-1H-indol-1-yl)-1,1,1-trifluoropropan-2-ol, tech

Lieferant: Apollo Scientific

PC28123-1G
APOSPC28123-1GEA 1570 EUR
APOSPC28123-1G
3-(2,3-Dihydro-1H-indol-1-yl)-1,1,1-trifluoropropan-2-ol, tech
3-(2,3-Dihydro-1H-indol-1-yl)-1,1,1-trifluoropropan-2-ol
Formel: C₁₁H₁₂F₃NO
Molecular Weight: 231,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02083039
CAS-Nummer: 672952-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR