3-(1-Azepanyl)-1,1,1-trifluoro-2-propanol 95%

Lieferant: Apollo Scientific

PC28124-1G
APOSPC28124-1GEA 894 EUR
APOSPC28124-1G
3-(1-Azepanyl)-1,1,1-trifluoro-2-propanol 95%
3-(1-Azepanyl)-1,1,1-trifluoro-2-propanol
Formel: C₉H₁₆F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD02083040
CAS-Nummer: 672952-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR