2-Hydroxy-3-phenyl-6-(trifluoromethyl)-3,4-dihydropyrimidin-4-one, tech
Lieferant: Apollo Scientific
PC28127-1G
APOSPC28127-1GEA
1570
EUR
APOSPC28127-1G
2-Hydroxy-3-phenyl-6-(trifluoromethyl)-3,4-dihydropyrimidin-4-one, tech
2-Hydroxy-3-phenyl-6-(trifluoromethyl)-3,4-dihydropyrimidin-4-one
Formel:
C₁₁H₇F₃N₂O₂ Molecular Weight: 256,181 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03102547 CAS-Nummer: 26676-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...