1-[2-Amino-4,6-bis(trifluoromethyl)phenyl]ethan-1-one, tech
Lieferant: Apollo Scientific
PC28128-1G
APOSPC28128-1GEA
1570
EUR
APOSPC28128-1G
1-[2-Amino-4,6-bis(trifluoromethyl)phenyl]ethan-1-one, tech
1-[2-Amino-4,6-bis(trifluoromethyl)phenyl]ethan-1-one
Formel:
C₁₀H₇F₆NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08281871 CAS-Nummer: 882747-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...