2-(4-Fluorophenyl)-7-methylimidazo[1,2-a]pyrimidin-5-ol, tech

Lieferant: Apollo Scientific

PC28130-1G
APOSPC28130-1GEA 1570 EUR
APOSPC28130-1G
2-(4-Fluorophenyl)-7-methylimidazo[1,2-a]pyrimidin-5-ol, tech
2-(4-Fluorophenyl)-7-methylimidazo[1,2-a]pyrimidin-5-ol
Formel: C₁₃H₁₀FN₃O
Molecular Weight: 243.236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125870
CAS-Nummer: 866050-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR