2-{[(2-Fluorophenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione, tech

Lieferant: Apollo Scientific

PC28132-1G
APOSPC28132-1GEA 1570 EUR
APOSPC28132-1G
2-{[(2-Fluorophenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione, tech
2-{[(2-Fluorophenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione
Formel: C₁₅H₁₆FNO₂
Molecular Weight: 261,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00816217
CAS-Nummer: 370853-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR