1,1,1-Trifluoro-3-(4-methylpiperidin-1-yl)propan-2-ol, tech
Lieferant: Apollo Scientific
PC28134-1G
APOSPC28134-1GEA
1570
EUR
APOSPC28134-1G
1,1,1-Trifluoro-3-(4-methylpiperidin-1-yl)propan-2-ol, tech
1,1,1-Trifluoro-3-(4-methylpiperidin-1-yl)propan-2-ol
Formel:
C₉H₁₆F₃NO Molecular Weight: 211,225 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05256215 CAS-Nummer: 866051-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...