Methyl 1-[(2-chloro-6-fluorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate, tech

Lieferant: Apollo Scientific

PC28135-1G
APOSPC28135-1GEA 1570 EUR
APOSPC28135-1G
Methyl 1-[(2-chloro-6-fluorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate, tech
Methyl 1-[(2-chloro-6-fluorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate
Formel: C₁₄H₁₁ClFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00232689
CAS-Nummer: 400086-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR