1-[(2-Chloro-6-fluorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid, tech

Lieferant: Apollo Scientific

PC28136-1G
APOSPC28136-1GEA 1570 EUR
APOSPC28136-1G
1-[(2-Chloro-6-fluorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid, tech
1-[(2-Chloro-6-fluorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid
Formel: C₁₃H₉ClFNO₃
Molecular Weight: 281,667 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140086
CAS-Nummer: 339008-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR