4-Fluoro-3-oxo-1-phenyl-2,3-dihydro-1H-indene-1-carboxylic acid, tech

Lieferant: Apollo Scientific

PC28137-1G
APOSPC28137-1GEA 1570 EUR
APOSPC28137-1G
4-Fluoro-3-oxo-1-phenyl-2,3-dihydro-1H-indene-1-carboxylic acid, tech
4-Fluoro-3-oxo-1-phenyl-2,3-dihydro-1H-indene-1-carboxylic acid
Formel: C₁₆H₁₁FO₃
Molecular Weight: 270,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00232260
CAS-Nummer: 339010-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR