(3E)-2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(methoxyimino)propanenitrile, tech

Lieferant: Apollo Scientific

PC28139-1G
APOSPC28139-1GEA 1570 EUR
APOSPC28139-1G
(3E)-2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(methoxyimino)propanenitrile, tech
(3E)-2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(methoxyimino)propanenitrile
Formel: C₁₀H₇ClF₃N₃O
Molecular Weight: 277,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663744
CAS-Nummer: 344263-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR