4-(4-Fluorophenyl)-6-(3-methoxyphenyl)-2,3,4,5-tetrahydropyridazin-3-one, tech
Lieferant: Apollo Scientific
PC28141-1G
APOSPC28141-1GEA
1570
EUR
APOSPC28141-1G
4-(4-Fluorophenyl)-6-(3-methoxyphenyl)-2,3,4,5-tetrahydropyridazin-3-one, tech
4-(4-Fluorophenyl)-6-(3-methoxyphenyl)-2,3,4,5-tetrahydropyridazin-3-one
Formel:
C₁₇H₁₅FN₂O₂ Molecular Weight: 298,312 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00664434 CAS-Nummer: 344282-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...