1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl](methyl)amino}pyrrolidin-2-one, tech

Lieferant: Apollo Scientific

PC28143-1G
APOSPC28143-1GEA 1570 EUR
APOSPC28143-1G
1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl](methyl)amino}pyrrolidin-2-one, tech
1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl](methyl)amino}pyrrolidin-2-one
Formel: C₁₁H₁₁ClF₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00664606
CAS-Nummer: 339014-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR