1,1,1-Trifluoro-3-(thiomorpholin-4-yl)propan-2-ol, tech

Lieferant: Apollo Scientific

PC28145-1G
APOSPC28145-1GEA 1570 EUR
APOSPC28145-1G
1,1,1-Trifluoro-3-(thiomorpholin-4-yl)propan-2-ol, tech
1,1,1-Trifluoro-3-(thiomorpholin-4-yl)propan-2-ol
Formel: C₇H₁₂F₃NOS
Molecular Weight: 215,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871641
CAS-Nummer: 478068-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR