Methyl 4-(4-fluorobenzoyl)-1H-pyrrole-2-carboxylate, tech

Lieferant: Apollo Scientific

PC28146-1G
APOSPC28146-1GEA 1570 EUR
APOSPC28146-1G
Methyl 4-(4-fluorobenzoyl)-1H-pyrrole-2-carboxylate, tech
Methyl 4-(4-fluorobenzoyl)-1H-pyrrole-2-carboxylate
Formel: C₁₃H₁₀FNO₃
Molecular Weight: 247,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571424
CAS-Nummer: 478078-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR