2-Chloro-2,2-difluoro-1-(1H-indol-3-yl)ethan-1-ol, tech
Lieferant: Apollo Scientific
PC28147-1G
APOSPC28147-1GEA
1570
EUR
APOSPC28147-1G
2-Chloro-2,2-difluoro-1-(1H-indol-3-yl)ethan-1-ol, tech
2-Chloro-2,2-difluoro-1-(1H-indol-3-yl)ethan-1-ol
Formel:
C₁₀H₈ClF₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD02571237 CAS-Nummer: 478079-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...