(2R)-3-{1,4-Dioxa-8-azaspiro[4.5]decan-8-yl}-1,1,1-trifluoropropan-2-ol, tech

Lieferant: Apollo Scientific

PC28149-1G
APOSPC28149-1GEA 1570 EUR
APOSPC28149-1G
(2R)-3-{1,4-Dioxa-8-azaspiro[4.5]decan-8-yl}-1,1,1-trifluoropropan-2-ol, tech
(2R)-3-{1,4-Dioxa-8-azaspiro[4.5]decan-8-yl}-1,1,1-trifluoropropan-2-ol
Formel: C₁₀H₁₆F₃NO₃
Molecular Weight: 255,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06809966
CAS-Nummer: 477762-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR