2-Chloro-2,2-difluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-ol, tech
Lieferant: Apollo Scientific
PC28150-1G
APOSPC28150-1GEA
1570
EUR
APOSPC28150-1G
2-Chloro-2,2-difluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-ol, tech
2-Chloro-2,2-difluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-ol
Formel:
C₁₁H₁₀ClF₂NO Molecular Weight: 245,653 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02571244 CAS-Nummer: 478079-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...