2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]ethan-1-ol, tech
Lieferant: Apollo Scientific
PC28151-1G
APOSPC28151-1GEA
1570
EUR
APOSPC28151-1G
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]ethan-1-ol, tech
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]ethan-1-ol
Formel:
C₁₀H₉F₃N₂O Molecular Weight: 230,186 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01017153 CAS-Nummer: 116040-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...