1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]butane-1,3-dione, tech

Lieferant: Apollo Scientific

PC28152-1G
APOSPC28152-1GEA 1570 EUR
APOSPC28152-1G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]butane-1,3-dione, tech
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]butane-1,3-dione
Formel: C₁₀H₇ClF₃NO₂
Molecular Weight: 265,616 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125986
CAS-Nummer: 866132-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR