5-Ethoxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile, tech
Lieferant: Apollo Scientific
PC28153-1G
APOSPC28153-1GEA
1040
EUR
APOSPC28153-1G
5-Ethoxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile, tech
5-Ethoxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile
Formel:
C₈H₈F₃N₃O Molecular Weight: 219,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04125990 CAS-Nummer: 955965-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...